ENA308584106 Enamine

2-(thiophen-2-yl)-1,3-thiazole-4-carbaldehyde

  • MDL number MFCD12776030
  • Cas Number : 256420-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68974

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68974
Availability details for
Quantity
  • Availability
  • Add to Cart