ENA308584109 Enamine

5-(4-methanesulfonylphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09815338
  • Cas Number : 1016504-27-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68977

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68977
Availability details for
Quantity
  • Availability
  • Add to Cart