ENA308584110 Enamine

2-methoxypyridine-3-carbothioamide

  • MDL number MFCD09947360
  • Cas Number : 1016837-97-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-68978
Availability details for
Quantity
  • Availability
  • Add to Cart