ENA308584111 Enamine

5-(5-chloro-2-methoxyphenyl)-1,3,4-oxadiazol-2-amine

  • MDL number MFCD09807184
  • Cas Number : 1016686-46-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 201...203
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68980

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68980
Availability details for
Quantity
  • Availability
  • Add to Cart