ENA308584112 Enamine

1-chloro-3-(propan-2-yloxy)propane

  • MDL number MFCD09753651
  • Cas Number : 72468-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-68981

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P363,P501
hcodes Xi,Xn
MSDS https://www.enaminestore.com/catalog/EN300-68981
pictograms GHS05,GHS07
hstatements H302,H312,H318,H332
Availability details for
Quantity
  • Availability
  • Add to Cart