ENA308584119 Enamine

{4-[3-(dimethylamino)propoxy]phenyl}methanamine

  • MDL number MFCD07786701
  • Cas Number : 91637-76-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-68989

Safety Information

msds https://www.enaminestore.com/catalog/EN300-68989
Availability details for
Quantity
  • Availability
  • Add to Cart