ENA308584126 Enamine

2-(4-bromophenyl)-2-(piperazin-1-yl)acetonitrile

  • MDL number MFCD09816843
  • Cas Number : 1000406-20-1
  • Molecular Weight : 280.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69007

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69007
Availability details for
Quantity
  • Availability
  • Add to Cart