ENA308584140 Enamine

2-methyl-2-[3-(propan-2-yl)phenoxy]propanoic acid

  • MDL number MFCD09712151
  • Cas Number : 857351-97-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69022

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69022
Availability details for
Quantity
  • Availability
  • Add to Cart