ENA308584142 Enamine

methyl 2-[1-(3-hydroxyphenyl)-N-methylformamido]acetate

  • MDL number MFCD11148064
  • Cas Number : 1020933-40-7
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69024

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69024
Availability details for
Quantity
  • Availability
  • Add to Cart