ENA308584151 Enamine

4-(1,3-thiazol-2-yl)piperazine-1-carbothioamide

  • MDL number MFCD09811650
  • Cas Number : 1016764-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69037

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69037
Availability details for
Quantity
  • Availability
  • Add to Cart