ENA308584158 Enamine

1-[(piperidin-3-yl)carbonyl]azepane hydrochloride

  • MDL number MFCD13562208
  • Cas Number : 1220039-05-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157...159
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69045

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69045
Availability details for
Quantity
  • Availability
  • Add to Cart