ENA308584363 Enamine

2-(1,4-diazepan-1-yl)benzonitrile hydrochloride

  • MDL number MFCD17167246
  • Cas Number : 1258650-73-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69066

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69066
Availability details for
Quantity
  • Availability
  • Add to Cart