ENA308584365 Enamine

methyl N-(3-methylfuran-2-yl)carbamate

  • MDL number MFCD17480365
  • Cas Number : 1258649-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69073

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69073
Availability details for
Quantity
  • Availability
  • Add to Cart