ENA308584369 Enamine

2-{4H,5H,7H-thieno[2,3-c]pyran-7-yl}acetic acid

  • MDL number MFCD17167248
  • Cas Number : 1258649-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69122

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69122
Availability details for
Quantity
  • Availability
  • Add to Cart