ENA308584372 Enamine

3-(4-bromophenyl)-5-methyl-1H-1,2,4-triazole

  • MDL number MFCD17167250
  • Cas Number : 170230-86-1
  • Molecular Weight : 238.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69145

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69145
Availability details for
Quantity
  • Availability
  • Add to Cart