ENA308584374 Enamine

6-(1-aminoethyl)-1,2,3,4-tetrahydroquinolin-2-one hydrochloride

  • MDL number MFCD17167251
  • Cas Number : 1258650-15-5
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69173

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69173
Availability details for
Quantity
  • Availability
  • Add to Cart