ENA308584376 Enamine

[(3-amino-1,1,1-trifluoropropan-2-yl)oxy]cyclopentane hydrochloride

  • MDL number MFCD17167252
  • Cas Number : 1258650-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69196

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69196
Availability details for
Quantity
  • Availability
  • Add to Cart