ENA308584378 Enamine

2,8,14-triazatricyclo[8.4.0.0,2,6]tetradeca-1(10),11,13-trien-7-one

  • MDL number MFCD17480376
  • Cas Number : 1258639-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69214
Availability details for
Quantity
  • Availability
  • Add to Cart