ENA308584379 Enamine

5-(chloromethyl)-4H,7H-pyrazolo[1,5-a]pyrimidin-7-one

  • MDL number MFCD17480377
  • Cas Number : 1258651-02-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69224
Availability details for
Quantity
  • Availability
  • Add to Cart