ENA308584383 Enamine

3-{2,4,6-trimethyl-3-oxo-1H,2H,3H-pyrazolo[3,4-b]pyridin-5-yl}propanoic acid

  • MDL number MFCD17480382
  • Cas Number : 1258641-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69251

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69251
Availability details for
Quantity
  • Availability
  • Add to Cart