ENA308584389 Enamine

5,6-dihydro-1,4-dioxin-2-ylmethanol

  • MDL number MFCD17480388
  • Cas Number : 94105-19-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69294

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69294
Availability details for
Quantity
  • Availability
  • Add to Cart