ENA308584396 Enamine

5-(aminomethyl)-4-ethyl-4H-1,2,4-triazole-3-thiol hydrochloride

  • MDL number MFCD17480395
  • Cas Number : 1258649-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69356

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69356
Availability details for
Quantity
  • Availability
  • Add to Cart