ENA308584403 Enamine

N-(2H-1,3-benzodioxol-5-ylmethyl)-1-methylcyclopentan-1-amine hydrochloride

  • MDL number MFCD17480402
  • Cas Number : 1258650-28-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 219...221
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69387

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69387
Availability details for
Quantity
  • Availability
  • Add to Cart