ENA308584405 Enamine

4-benzyl-3,3-dimethylpiperazin-2-one

  • MDL number MFCD02930378
  • Cas Number : 29906-46-5
  • Molecular Weight : 218.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69395

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69395
Availability details for
Quantity
  • Availability
  • Add to Cart