ENA308584406 Enamine

2-methyl-4-[(propan-2-yl)amino]benzamide

  • MDL number MFCD17480404
  • Cas Number : 1258651-32-9
  • Molecular Weight : 192.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69407

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69407
Availability details for
Quantity
  • Availability
  • Add to Cart