ENA308584407 Enamine

4-chloro-2-methyl-1H-1,3-benzodiazol-1-ol

  • MDL number MFCD17480405
  • Cas Number : 1258649-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69410

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69410
Availability details for
Quantity
  • Availability
  • Add to Cart