ENA308584408 Enamine

methyl 3-amino-2-[(4-hydroxyphenyl)methyl]propanoate hydrochloride

  • MDL number MFCD17480406
  • Cas Number : 1258640-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 163...165
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69412

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69412
Availability details for
Quantity
  • Availability
  • Add to Cart