ENA308584409 Enamine

N-(5-cyano-2-methanesulfonylphenyl)acetamide

  • MDL number MFCD17480407
  • Cas Number : 1258649-82-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69414

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69414
Availability details for
Quantity
  • Availability
  • Add to Cart