ENA308584411 Enamine

2-(2-chloropyridin-4-yl)-6-methyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD17480409
  • Cas Number : 1258640-22-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 269...271
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69424

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69424
Availability details for
Quantity
  • Availability
  • Add to Cart