ENA308584412 Enamine

[2-(1H-1,3-benzodiazol-1-yl)pyridin-3-yl]methanamine dihydrochloride

  • MDL number MFCD17480411
  • Cas Number : 1258650-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Product Link https://www.enaminestore.com/catalog/EN300-69431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69431
Availability details for
Quantity
  • Availability
  • Add to Cart