ENA308584413 Enamine

4-aminocyclohexane-1-carboxamide hydrochloride

  • MDL number MFCD17480412
  • Cas Number : 856563-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 279...281
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69435

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69435
Availability details for
Quantity
  • Availability
  • Add to Cart