ENA308584419 Enamine

7-sulfamoyl-2,3-dihydro-1H-indene-5-carboxylic acid

  • MDL number MFCD17480418
  • Cas Number : 1258649-99-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69477

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69477
Availability details for
Quantity
  • Availability
  • Add to Cart