ENA308584420 Enamine

2-(chloromethyl)-5-methylpyrazine hydrochloride

  • MDL number MFCD17480419
  • Cas Number : 128229-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69482

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69482
Availability details for
Quantity
  • Availability
  • Add to Cart