ENA308584421 Enamine

3-(1-aminopropyl)benzene-1-sulfonamide hydrochloride

  • MDL number MFCD17480420
  • Cas Number : 889856-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-69489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69489
Availability details for
Quantity
  • Availability
  • Add to Cart