ENA308584422 Enamine

3-[methyl(3-phenoxypropyl)amino]propanoic acid hydrochloride

  • MDL number MFCD17480421
  • Cas Number : 132424-09-0
  • Molecular Weight : 273.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69494
Availability details for
Quantity
  • Availability
  • Add to Cart