ENA308584424 Enamine

5-chloro-4-acetamido-2-fluorobenzene-1-sulfonyl chloride

  • MDL number MFCD17480424
  • Cas Number : 1258651-40-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69518

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69518
Availability details for
Quantity
  • Availability
  • Add to Cart