ENA308584435 Enamine

(3-phenoxypropyl)hydrazine hydrochloride

  • MDL number MFCD17480438
  • Cas Number : 81866-67-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-69639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69639
Availability details for
Quantity
  • Availability
  • Add to Cart