ENA308584437 Enamine

[1-(methylamino)cyclohexyl]methanol hydrochloride

  • MDL number MFCD17480440
  • Cas Number : 1258651-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 166...168
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69654

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69654
Availability details for
Quantity
  • Availability
  • Add to Cart