ENA308584447 Enamine

2-(2-chloropyridin-4-yl)-6-ethyl-3,4-dihydropyrimidin-4-one

  • MDL number MFCD17480451
  • Cas Number : 1258651-71-6
  • Molecular Weight : 235.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69710

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69710
Availability details for
Quantity
  • Availability
  • Add to Cart