ENA308584452 Enamine

2-(2,6-dichlorophenyl)-2-methylpropan-1-amine hydrochloride

  • MDL number MFCD17480457
  • Cas Number : 959140-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69776
Availability details for
Quantity
  • Availability
  • Add to Cart