ENA308584455 Enamine

6-(azepan-1-yl)pyridine-3-carboxylic acid hydrochloride

  • MDL number MFCD17480460
  • Cas Number : 1258640-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-69846

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69846
Availability details for
Quantity
  • Availability
  • Add to Cart