ENA308584462 Enamine

2-phenyl-4-(piperazin-1-yl)pyrimidine dihydrobromide

  • MDL number MFCD17480469
  • Cas Number : 1258640-39-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HBr
Product Link https://www.enaminestore.com/catalog/EN300-69907

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69907
Availability details for
Quantity
  • Availability
  • Add to Cart