ENA308584560 Enamine

3-phenylpent-2-enoic acid

  • Cas Number : 174660-87-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-69307

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69307
Availability details for
Quantity
  • Availability
  • Add to Cart