ENA308584567 Enamine

5-(cyclohexylmethyl)-1,3,4-thiadiazol-2-amine

  • MDL number MFCD02664067
  • Cas Number : 118314-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69318

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69318
Availability details for
Quantity
  • Availability
  • Add to Cart