ENA308584569 Enamine

3-hydroxy-4-methylbenzamide

  • MDL number MFCD11110232
  • Cas Number : 135679-95-7
  • Molecular Weight : 151.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69321

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69321
Availability details for
Quantity
  • Availability
  • Add to Cart