ENA308584570 Enamine

(2,6-dichlorophenyl)methanesulfonyl chloride

  • MDL number MFCD11204033
  • Cas Number : 85952-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 70...72
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69322

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69322
Availability details for
Quantity
  • Availability
  • Add to Cart