ENA308584576 Enamine

methyl[(1-methylpiperidin-4-yl)methyl]amine

  • MDL number MFCD06738983
  • Cas Number : 405928-19-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69329

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69329
Availability details for
Quantity
  • Availability
  • Add to Cart