ENA308584581 Enamine

ethyl[(4-fluoro-3-methylphenyl)(pyridin-3-yl)methyl]amine

  • MDL number MFCD12877800
  • Cas Number : 1179861-37-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69334

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69334
Availability details for
Quantity
  • Availability
  • Add to Cart