ENA308584587 Enamine

N-methyl-1-[(3-methyl-1,2-oxazol-5-yl)methyl]piperidin-4-amine

  • MDL number MFCD16125972
  • Cas Number : 1251099-46-3
  • Molecular Weight : 209.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-69086

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-69086
Availability details for
Quantity
  • Availability
  • Add to Cart