ENA308584589 Enamine

6-(2,2,3,3-tetrafluoropropoxy)pyridin-3-amine

  • MDL number MFCD11197493
  • Cas Number : 915394-36-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-69345

Safety Information

msds https://www.enaminestore.com/catalog/EN300-69345
Availability details for
Quantity
  • Availability
  • Add to Cart